Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, 99.8%, for HPLC

Catalog No. AC433580010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433580010 1 L Glass Bottle
AC433580025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433580010 Supplier Thermo Scientific Chemicals Supplier No. 433580010
Add to Cart

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35.0°C
Absorbance 0.005 max. at 260nm, 0.005 max. at 270nm, 0.01 max. at 300nm, 0.01 max. at 400nm, 0.05 max. at 250nm, 0.15 max. at 240nm, 1 max. at 230nm
Color White
Acidity 0.0001 meq/g max.
Alkalinity 0.0001 meq/g max.
Density 1.2500g/mL
Quantity 1 L
Flash Point 13°C
Packaging Glass Bottle
Refractive Index 1.4440 to 1.4460
Residue after Evaporation 2ppm max.
Assay Percent Range 99.75% min. (GC)
Specific Gravity 1.25
Linear Formula ClCH2CH2Cl
Viscosity 0.8 mPa.s (20°C)
Water 0.02% Max.
Merck Index 15,3851
Boiling Point 81.0°C to 85.0°C
Solubility Information Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 98.96
Percent Purity 99.80%
Grade HPLC
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wa
EINECSNumber 203-458-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.