Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, 99+%, for spectroscopy

Catalog No. AC167765000
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167765000 500 mL Glass Bottle
AC167760010 1 L Glass Bottle
AC167760025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167765000 Supplier Thermo Scientific Chemicals Supplier No. 167765000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35.0°C
Absorbance 0.01 max. at 300nm, 0.15 max. at 250nm, 1 max. at 230nm
CAS Min % 99.0
CAS Max % 100.0
Density 1.2500g/mL
Quantity 500 mL
Flash Point 13°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4440 to 1.4460
Residue after Evaporation 0.002% max.
Specific Gravity 1.25
Assay Percent Range 99+%
Linear Formula ClCH2CH2Cl
Viscosity 0.8 mPa.s (20°C)
Water 0.03% max.
Beilstein 01, 84
Merck Index 15, 3851
Boiling Point 81.0°C to 85.0°C
Solubility Information Solubility in water: 8.7g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 98.96
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material 1,2-Dichloroethane, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-458-1
RTECSNumber KI0525000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.