Learn More
1,2-Dichloroethane-d{4}, 99% (Isotopic)
Description
1,2-Dichloroethane-d4 is widely used as a solvent for NMR analysis, and in the synthesis of poly(ethylene-d4 oxide) as a chain termination reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 5 g |
| Refractive Index | 1.443 |
| Density | 1.307 g/mL |
| Flash Point | 15°C (60°F) |
| Melting Point | -35°C |
| Boiling Point | 83°C |
| UN Number | UN1184 |
| Solubility Information | Partially miscible with water. |
| Formula Weight | 102.99 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H315-H319-H335-H350 |
| Hazard Statement | GHS H Statement H225-H350-H302-H315-H319-H335 Highly flammable liquid and vapor. May cause cancer. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE |
| TSCA | No |
| Recommended Storage | Ambient |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.