Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, puriss. p.a., ≥99.5% (GC), Solstice

Catalog No. 60044520 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-520 2.5 L Glass Bottle
60-044-519 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-044-520 Supplier Solstice Supplier No. 035302.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point ∼ -35°C
Color Colorless
Density 1.256g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.156
Flash Point 13°C (55.4°F)
Ignition Point 775°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 87mmHg (25°C)
Linear Formula ClCH2CH2Cl
UN Number UN1184
Beilstein 605264
Boiling Point ∼83°C
Formula Weight 98.96g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
Hazard Statement P201-P210-P261-P305 + P351 + P338-P311
Precautionary Statement H225-H302-H315-H319-H331-H335-H350
DOTInformation Class 3 - PG 2 - Ethylene dichloride
EINECSNumber 203-458-1
RTECSNumber KI0525000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.