Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences 1,2-Dichloroethane, Reagent, A.C.S. 450 ML
SDP

Catalog No. 50980358
Encompass_Preferred

CAS #107-06-2 CH₂ClCH₂Cl F.W. 98.96
Solubility: Ethylene Dichloride
Boiling point: 20°C

This item is not returnable. View return policy
Catalog No. 50-980-358 Supplier Electron Microscopy Sciences Supplier No. 13250
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #107-06-2 CH₂ClCH₂Cl F.W. 98.96
Solubility: Ethylene Dichloride
Boiling point: 20°C

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.