Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences 1,2-Dichloroethane, Reagent, A.C.S. 450 ML
SDP

Supplier:  Electron Microscopy Sciences 13251

Encompass_Preferred

CAS #107-06-2 CH₂ClCH₂Cl F.W. 98.96
Solubility: Ethylene Dichloride
Boiling point: 20°C

Catalog No. 50-980-359


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.