Learn More
Enzo Life Sciences (±)1,2-Didecanoylglycerol (10:0) (10mg). CAS: 17863-69-3
SDP
Catalog No. 502010505
Encompass_Preferred
A membrane permeable but non-physiologic DAG. Stimulates protein kinase C in intact platelets. Purity: ≥98% (TLC). Long Term Storage: -20°C.
Catalog No. 50-201-0505 Supplier Enzo Life Sciences Supplier No. BMLDG1000010
A membrane permeable but non-physiologic DAG. Stimulates protein kinase C in intact platelets. Purity: ≥98% (TLC). Long Term Storage: -20°C.