Learn More
Enzo Life Sciences (±)1,2-Didecanoylglycerol (10:0) (50mg). CAS: 17863-69-3
SDP
Supplier:  Enzo Life Sciences BMLDG1000050
Encompass_Preferred
A membrane permeable but non-physiologic DAG. Stimulates protein kinase C in intact platelets. Purity: ≥98% (TLC). Long Term Storage: -20°C.
Catalog No. 50-201-0506