Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diethoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D04525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04525G 5 g
D045225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04525G Supplier TCI America Supplier No. D04525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2050-46-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015143
InChI Key QZYDOKBVZJLQCK-UHFFFAOYSA-N
Synonym o-diethoxybenzene, benzene, 1,2-diethoxy, catechol diethyl ether, benzene, o-diethoxy, o-diethoxy benzene, benzene, diethoxy, unii-p9dm5t78pz, pyrocatechol diethyl ether, p9dm5t78pz, diethoxybenzene-1,2
PubChem CID 74904
IUPAC Name 1,2-diethoxybenzene
SMILES CCOC1=CC=CC=C1OCC

Specifications

Melting Point 44°C
Color White
Boiling Point 220°C
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Diethoxybenzene
RTECSNumber CZ5580000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.