Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2-(diethylamino)ethyl)-3-(2-(4-ethylpiperazin-1-yl)-4-methylquinolin-6-yl)thiourea | 688342-78-1 | 99.7% | 428.64 | C23H36N6S | 1MG
SDP

Catalog No. 5000237171
Encompass_Preferred

D-I03 is a small-molecule selective RAD52 inhibitor used as a research reagent to study RAD52-dependent single-strand annealing and D-loop formation. It shows cellular activity against BRCA1- and BRCA2-deficient cells.

  • Selective RAD52 inhibitor with Kd 25.8 μM.
  • Inhibits RAD52-dependent single-strand annealing and D-loop formation.
  • Cellular activity: suppresses growth of BRCA1- and BRCA2-deficient cells.
  • Reported IC50 values: 5 μM (SSA) and 8 μM (D-loop).
  • Purity 99.7%.
  • Molecular weight 428.64 g/mol; formula C23H36N6S.
  • Physical state solid; color off-white to light yellow.
  • Solubility in DMSO 50 mg/mL.

Catalog No. 50-002-37171 Supplier Medchemexpress LLC Supplier No. HY1246911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-I03 is a small-molecule selective RAD52 inhibitor used as a research reagent to study RAD52-dependent single-strand annealing and D-loop formation. It shows cellular activity against BRCA1- and BRCA2-deficient cells.

  • Selective RAD52 inhibitor with Kd 25.8 μM.
  • Inhibits RAD52-dependent single-strand annealing and D-loop formation.
  • Cellular activity: suppresses growth of BRCA1- and BRCA2-deficient cells.
  • Reported IC50 values: 5 μM (SSA) and 8 μM (D-loop).
  • Purity 99.7%.
  • Molecular weight 428.64 g/mol; formula C23H36N6S.
  • Physical state solid; color off-white to light yellow.
  • Solubility in DMSO 50 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.