Alfa Aesar™ 1,2-Difluoro-4-iodobenzene, 99%

Manufacturer: Alfa Aesar™ A1133906

Quantity 5g

 View more versions of this product

Catalog No. AAA1133906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64248-58-4
    Molecular Formula C6H3F2I
    Formula Weight 239.99
    MDL Number MFCD00044630
    Beilstein 2081099
    Synonym 3,4-Difluoroiodobenzene
    Chemical Name or Material 1,2-Difluoro-4-iodobenzene

    Specifications

    Formula Weight 239.99
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00044630
    Synonym 3,4-Difluoroiodobenzene
    Boiling Point 177° to 178°C
    Quantity 5g
    Molecular Formula C6H3F2I
    CAS 64248-58-4
    Beilstein 2081099
    Flash Point 72°C (161°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Chemical Name or Material 1,2-Difluoro-4-iodobenzene
    Assay 99%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit