Learn More
Medchemexpress LLC 1,2-diheptanoyl-sn-glycero-3-phosphocholine | 39036-04-9 | 99.5% | 250 MG
SDP
Catalog No. 5000231385
Encompass_Preferred
1,2-diheptanoyl-sn-glycero-3-phosphocholine is a synthetic short-chain phosphatidylcholine used as a membrane mimic and mild detergent in biochemical and biophysical research. It supports micelle and short-chain lipid assembly formation for studies of membrane proteins, lipid-protein interactions, and membrane dynamics.
  • Short-chain phospholipid suitable as a detergent and membrane mimic.
  • High purity for reproducible biochemical assays.
  • Soluble in common organic solvents for lipid formulation.
  • Facilitates consistent micelle and bicelle formation.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-31385 Supplier Medchemexpress LLC Supplier No. HY122056250MG
1,2-diheptanoyl-sn-glycero-3-phosphocholine is a synthetic short-chain phosphatidylcholine used as a membrane mimic and mild detergent in biochemical and biophysical research. It supports micelle and short-chain lipid assembly formation for studies of membrane proteins, lipid-protein interactions, and membrane dynamics.
  • Short-chain phospholipid suitable as a detergent and membrane mimic.
  • High purity for reproducible biochemical assays.
  • Soluble in common organic solvents for lipid formulation.
  • Facilitates consistent micelle and bicelle formation.
  • Stable when stored under recommended conditions.