Learn More
AdipoGen 1 2-Dihydro-naphthalin
SDP
Supplier:  AdipoGen CDXD1139G005
Encompass_Preferred
Chemical. CAS 447-53-0. Formula C10H10. MW 130.19. 1,2-Dihydronaphthalene can be used as a building block, starting material or reagent in organic synthesis to create more complex chemical compounds and fluorescent dyes.
Catalog No. 50-256-1013