Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dihydronaphthalene 98.0+%, TCI America™
SDP

Catalog No. D05851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05851G 1 g
D058510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05851G Supplier TCI America Supplier No. D05851G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 447-53-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00001672
InChI Key KEIFWROAQVVDBN-UHFFFAOYSA-N
PubChem CID 9938
ChEBI CHEBI:38142
IUPAC Name 1,2-dihydronaphthalene
SMILES C1CC2=CC=CC=C2C=C1

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 207°C
Quantity 1 g
Synonym dialin, 1,2-dialin, diolin, delta1-dialin, dihydronaphthalene, naphthalene, 1,2-dihydro, dialin van, delta 1-dialin, dihydronapthalene, naphthalene, dihydro
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dihydronaphthalene
EINECSNumber (4)-1177
RTECSNumber QJ4375000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.