Learn More
Medchemexpress LLC 1,2-Dihydroxynaphthalene | 574-00-5 | 98.9% | 250 MG
SDP
Catalog No. 5000342478
Encompass_Preferred
1,2-Dihydroxynaphthalene is an intermediate isolated from the metabolism of naphthalene and has been shown to be a precursor to the formation of naphthoquinones. It can be further metabolized by some bacteria (such as Pseudomonas) to compounds such as coumarins and salicylic acid.
  • Appearance: solid
  • Color: off-white to light yellow
  • Structure classification: phenols, polyphenols
  • Initial source: microorganisms
Catalog No. 50-003-42478 Supplier Medchemexpress LLC Supplier No. 250MGHY128105
1,2-Dihydroxynaphthalene is an intermediate isolated from the metabolism of naphthalene and has been shown to be a precursor to the formation of naphthoquinones. It can be further metabolized by some bacteria (such as Pseudomonas) to compounds such as coumarins and salicylic acid.
  • Appearance: solid
  • Color: off-white to light yellow
  • Structure classification: phenols, polyphenols
  • Initial source: microorganisms