Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Diiodobenzene

Catalog No. 2381121G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-811-21G 1 g
23-811-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-811-21G Supplier Sigma Aldrich Supplier No. 2381121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Photolysis of 1,2-diiodobenzene in cyclohexane, furan, benzene and benzene containing tetraphenylcyclopentadienone has been studied.1,2-Diiodobenzene has been used in the preparation of purino[8,9-f]phenanthridines.

Specifications

CAS 615-42-9
Density 2.524 g/mL (at 25°C (literature))
Boiling Point 152°C (15 mmHg)
Molecular Formula C6H4I2
Refractive Index n20/D 1.718 (literature)
Linear Formula C6H4I2
MDL Number MFCD00001038
Quantity 1 g
Molecular Weight (g/mol) 329.9
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.