Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diiodotetrafluoroethane, 96%, stab. with copper

Catalog No. AAA1095406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1095406 5 g
AAA1095403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1095406 Supplier Thermo Scientific Chemicals Supplier No. A1095406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 354-65-4 | C2F4I2 | 353.825 g/mol

1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-65-4
Molecular Formula C2F4I2
Molecular Weight (g/mol) 353.825
MDL Number MFCD00039401
InChI Key NZXVPCQHQVWOFD-UHFFFAOYSA-N
Synonym 1,2-diiodotetrafluoroethane, 1,2-diiodoperfluoroethane, unii-2z13i2welk, ethane, 1,1,2,2-tetrafluoro-1,2-diiodo, 2z13i2welk, acmc-1cknr
PubChem CID 67720
IUPAC Name 1,1,2,2-tetrafluoro-1,2-diiodoethane
SMILES C(C(F)(F)I)(F)(F)I

Specifications

Melting Point -21°C
Density 2.629
Boiling Point 112°C to 113°C
Flash Point None
Refractive Index 1.495
Quantity 5 g
Beilstein 1740350
Sensitivity Light sensitive
Formula Weight 353.82
Percent Purity 96%
Chemical Name or Material 1,2-Diiodotetrafluoroethane, Stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-567-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.