Learn More
Medchemexpress LLC 1,2-dilauroyl-3-chloropropanediol | 1051389-99-1 | MFCD34647394 | >98.0% | 475.14 g·mol⁻¹ | C27H51ClO4 | 5 MG
SDP
Catalog No. 5000285061
Encompass_Preferred
1,2-dilauroyl-3-chloropropanediol is a chloropropanediol ester used as a research-grade lipid reagent for analytical and synthetic applications. It has molecular formula C27H51ClO4, molecular weight ≈475.14 g·mol⁻¹, and CAS 1051389-99-1. The compound is typically supplied as a solid with reported purity >98% and is recommended to be stored at -20 °C.
  • High purity (>98%) suitable for analytical and synthetic workflows.
  • Well-defined molecular identity: C27H51ClO4; MW 475.14 g·mol⁻¹; CAS 1051389-99-1.
  • Solid form facilitates accurate weighing and handling.
  • Small pack sizes available for method development and reference use.
  • Recommended cold storage to maintain stability.
Catalog No. 50-002-85061 Supplier Medchemexpress LLC Supplier No. HYW5875075MG
1,2-dilauroyl-3-chloropropanediol is a chloropropanediol ester used as a research-grade lipid reagent for analytical and synthetic applications. It has molecular formula C27H51ClO4, molecular weight ≈475.14 g·mol⁻¹, and CAS 1051389-99-1. The compound is typically supplied as a solid with reported purity >98% and is recommended to be stored at -20 °C.
  • High purity (>98%) suitable for analytical and synthetic workflows.
  • Well-defined molecular identity: C27H51ClO4; MW 475.14 g·mol⁻¹; CAS 1051389-99-1.
  • Solid form facilitates accurate weighing and handling.
  • Small pack sizes available for method development and reference use.
  • Recommended cold storage to maintain stability.