Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dilauroyl-sn-glycero-3-phosphoethanolamine | 59752-57-7 | MFCD00042828 | 98.0% | 579.75 g·mol⁻1 | C29H58NO8P | 50 MG
SDP

Catalog No. 5000282515
Encompass_Preferred

1,2-Dilauroyl-sn-glycero-3-phosphoethanolamine is a phosphatidylethanolamine lipid with lauric (12:0) acyl chains. It is commonly used as a lipid component for preparing liposomes and for studies of membrane composition in biochemical and biophysical research.

  • High purity suitable for research applications.
  • Useful for liposome synthesis and membrane composition studies.
  • Available in multiple small pack sizes for laboratory use.
  • Contains lauric (12:0) acyl chains for defined lipid composition.
  • Molecular weight 579.75 g·mol⁻1 and formula C29H58NO8P for accurate formulation.

Catalog No. 50-002-82515 Supplier Medchemexpress LLC Supplier No. HYW32222550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dilauroyl-sn-glycero-3-phosphoethanolamine is a phosphatidylethanolamine lipid with lauric (12:0) acyl chains. It is commonly used as a lipid component for preparing liposomes and for studies of membrane composition in biochemical and biophysical research.

  • High purity suitable for research applications.
  • Useful for liposome synthesis and membrane composition studies.
  • Available in multiple small pack sizes for laboratory use.
  • Contains lauric (12:0) acyl chains for defined lipid composition.
  • Molecular weight 579.75 g·mol⁻1 and formula C29H58NO8P for accurate formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.