Learn More
Medchemexpress LLC 1,2-dilauroyl-sn-glycero-3-phosphoethanolamine | 59752-57-7 | MFCD00042828 | 98.0% | 579.75 g·mol⁻¹ | C29H58NO8P | 100 MG
SDP
Catalog No. 5000280790
Encompass_Preferred
1,2-Dilauroyl-sn-glycero-3-phosphoethanolamine (DLPE) is a phosphatidylethanolamine derivative bearing two lauroyl (C12:0) acyl chains. It is a research-grade lipid commonly used for liposome formulation and membrane model studies; typical properties include a molecular weight of 579.75 g·mol⁻¹ and high purity (98.0%).
  • Phosphatidylethanolamine lipid with two lauroyl (C12:0) chains.
  • Suitable for liposome synthesis and membrane model studies.
  • High purity (98.0%) for research applications.
  • Molecular formula C29H58NO8P and molecular weight 579.75 g·mol⁻¹.
Catalog No. 50-002-80790 Supplier Medchemexpress LLC Supplier No. HYW322225100MG
1,2-Dilauroyl-sn-glycero-3-phosphoethanolamine (DLPE) is a phosphatidylethanolamine derivative bearing two lauroyl (C12:0) acyl chains. It is a research-grade lipid commonly used for liposome formulation and membrane model studies; typical properties include a molecular weight of 579.75 g·mol⁻¹ and high purity (98.0%).
  • Phosphatidylethanolamine lipid with two lauroyl (C12:0) chains.
  • Suitable for liposome synthesis and membrane model studies.
  • High purity (98.0%) for research applications.
  • Molecular formula C29H58NO8P and molecular weight 579.75 g·mol⁻¹.