Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-Dilauroyl-sn-glycerol | 60562-15-4 | 98.0% | 50 MG
SDP

Catalog No. 5000340253
Encompass_Preferred

1,2-Dilauroyl-sn-glycerol is a saturated diacylglycerol that may be involved in second messenger signal transduction. It appears as a white to off-white solid with a molecular formula of C27H52O5 and a molecular weight of 456.70. This product is intended for research use only.

  • Purity of 98.0%
  • CAS number: 60562-15-4
  • Soluble in DMSO (100 mg/mL), requiring ultrasonic for dissolution
  • Ships at room temperature within the continental US
  • Recommended storage for powder: -20°C for 3 years
  • Recommended storage for in-solvent solutions: -80°C for 6 months, or -20°C for 1 month (aliquot and store to prevent inactivation from repeated freeze-thaw cycles)
  • Related to metabolic enzymes/proteases and endogenous metabolites
  • Acts as an inhibitor

Catalog No. 50-003-40253 Supplier Medchemexpress LLC Supplier No. HY12846550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dilauroyl-sn-glycerol is a saturated diacylglycerol that may be involved in second messenger signal transduction. It appears as a white to off-white solid with a molecular formula of C27H52O5 and a molecular weight of 456.70. This product is intended for research use only.

  • Purity of 98.0%
  • CAS number: 60562-15-4
  • Soluble in DMSO (100 mg/mL), requiring ultrasonic for dissolution
  • Ships at room temperature within the continental US
  • Recommended storage for powder: -20°C for 3 years
  • Recommended storage for in-solvent solutions: -80°C for 6 months, or -20°C for 1 month (aliquot and store to prevent inactivation from repeated freeze-thaw cycles)
  • Related to metabolic enzymes/proteases and endogenous metabolites
  • Acts as an inhibitor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.