Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Dimethoxy-1,1,2,2-Tetramethyldisilane

Catalog No. 6781631G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-816-31G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 67-816-31G Supplier Sigma Aldrich Supplier No. 6781631G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Dimethoxy-1,1,2,2-tetramethyldisilane can be used:In the palladium catalyzed bis(silylation) of 1,4-diethynylbenzene to obtain a crack free sol-gel.To prepare dimethylsilylene via gas-phase flow-pyrolysis at 600 0C.

Specifications

CAS 207124-65-0
Melting Point >300°C
Molecular Formula C17H19ClN2
Refractive Index n20/D 1.425
Linear Formula C17H19ClN2
Quantity 250 mg
Synonym 2-(2,4,6-Trimethylphenyl)-5-methylimidazo[1,5-a]pyridinim chloride
Molecular Weight (g/mol) 286.8 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.