Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,2-Dimethoxyethane, 110-71-4, MFCD00008502, 100 mL
Catalog No. NC0915846
Encompass
Linear Formula: CH3OCH2CH2OCH3, Molecular Weight: 90.12, anhydrous, 99.5%, inhibitor-free, Synonym: DME, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme, mono-Glyme. **For R&D use only. Not for drug, household, or other uses.**
This item is not returnable. View return policy
Catalog No. NC0915846 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 259527100ML
Linear Formula: CH3OCH2CH2OCH3, Molecular Weight: 90.12, anhydrous, 99.5%, inhibitor-free, Synonym: DME, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme, mono-Glyme. **For R&D use only. Not for drug, household, or other uses.**