Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethyl-3H-benzo[e]indole 98.0+%, TCI America™
SDP

Catalog No. D51635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51635G 5 g
D51631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51635G Supplier TCI America Supplier No. D51635G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55970-05-3
Molecular Formula C14H13N
MDL Number MFCD00850046

Specifications

Melting Point 132°C
Color Gray-Red
Quantity 5 g
Formula Weight 195.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethyl-3H-benzo[e]indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.