Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dimethyl-5-nitroimidazole-d3 | 64678-69-9 | MFCD09265282 | 99.9% | 144.15 | C5H4D3N3O2 | 10 MG
SDP

Catalog No. 5000297747
Encompass_Preferred

Dimetridazole-d3 is the deuterium-labeled analog of dimetridazole supplied as a solid analytical standard for research and quantitative analysis. It is used as an internal standard or isotopic tracer in mass spectrometry and other analytical workflows, providing well-defined isotopic composition and traceability.

  • Isotope-labeled internal standard for mass spectrometry.
  • High purity (≈99.9%) suitable for analytical use.
  • Molecular weight 144.15 and formula C5H4D3N3O2.
  • Provided as a 10 MG solid powder; other pack sizes available.
  • Powder storage: -20 °C (3 years) or 4 °C (2 years); in solvent: -80 °C (6 months).

Catalog No. 50-002-97747 Supplier Medchemexpress LLC Supplier No. HYB1244S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimetridazole-d3 is the deuterium-labeled analog of dimetridazole supplied as a solid analytical standard for research and quantitative analysis. It is used as an internal standard or isotopic tracer in mass spectrometry and other analytical workflows, providing well-defined isotopic composition and traceability.

  • Isotope-labeled internal standard for mass spectrometry.
  • High purity (≈99.9%) suitable for analytical use.
  • Molecular weight 144.15 and formula C5H4D3N3O2.
  • Provided as a 10 MG solid powder; other pack sizes available.
  • Powder storage: -20 °C (3 years) or 4 °C (2 years); in solvent: -80 °C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.