Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethyl-5-nitroimidazole 98.0+%, TCI America™
SDP

Catalog No. D156425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D156425G 25 g
D1564500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D156425G Supplier TCI America Supplier No. D156425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 551-92-8 | C5H7N3O2 | 141.13 g/mol

Chemical Identifiers

CAS 551-92-8
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
MDL Number MFCD00047046
InChI Key IBXPYPUJPLLOIN-UHFFFAOYSA-N
Synonym dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
PubChem CID 3090
IUPAC Name 1,2-dimethyl-5-nitro-1H-imidazole
SMILES CN1C(C)=NC=C1[N+]([O-])=O

Specifications

Melting Point 140°C
Color Yellow
Quantity 25 g
Formula Weight 141.13
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethyl-5-nitroimidazole
RTECSNumber NI5075000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.