Learn More
Medchemexpress LLC 1-(2-(dimethylamino)ethyl)-3-(7-methylbenzo[1,2-d:3,4-d']bis(thiazole)-2-yl)urea | 1428729-63-8 | MFCD31619259 | 99.5% | 335.45 | C14H17N5OS2 | 10 MG
SDP
Catalog No. 5000242844
Encompass_Preferred
RIG-1 modulator 1 is a small-molecule research compound described as an antiviral agent with reported activity against influenza virus, hepatitis B virus (HBV), hepatitis C virus (HCV), and human immunodeficiency virus (HIV). It is supplied as a solid for laboratory research; key analytical data include molecular formula C14H17N5OS2 and molecular weight 335.45 g/mol.
Catalog No. 50-002-42844 Supplier Medchemexpress LLC Supplier No. HY10790210MG
RIG-1 modulator 1 is a small-molecule research compound described as an antiviral agent with reported activity against influenza virus, hepatitis B virus (HBV), hepatitis C virus (HCV), and human immunodeficiency virus (HIV). It is supplied as a solid for laboratory research; key analytical data include molecular formula C14H17N5OS2 and molecular weight 335.45 g/mol.