Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2-(dimethylamino)ethyl)-3-(7-methylbenzo[1,2-d:3,4-d']bis(thiazole)-2-yl)urea | 1428729-63-8 | MFCD31619259 | 99.5% | 335.45 | C14H17N5OS2 | 100 MG
SDP

Catalog No. 5000242667
Encompass_Preferred

RIG-1 modulator 1 is a small-molecule antiviral research compound described as "Compound of claim 13" that modulates RIG-1-related activity and is reported active against influenza virus, HBV, HCV, and HIV. It is supplied as a characterized solid for in vitro research and includes storage and stability information for powder and solutions.

  • High purity: 99.5%.
  • Appearance: solid, white to light yellow.
  • Molecular formula: C14H17N5OS2; molecular weight: 335.45.
  • Available in multiple pack sizes, including 100 mg.
  • Storage recommendations: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solvent -80 °C (up to 2 years).
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-42667 Supplier Medchemexpress LLC Supplier No. HY107902100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RIG-1 modulator 1 is a small-molecule antiviral research compound described as "Compound of claim 13" that modulates RIG-1-related activity and is reported active against influenza virus, HBV, HCV, and HIV. It is supplied as a characterized solid for in vitro research and includes storage and stability information for powder and solutions.

  • High purity: 99.5%.
  • Appearance: solid, white to light yellow.
  • Molecular formula: C14H17N5OS2; molecular weight: 335.45.
  • Available in multiple pack sizes, including 100 mg.
  • Storage recommendations: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solvent -80 °C (up to 2 years).
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.