Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2-dimethylaminoethyl)-1,2,3,4-tetrahydro-2-methylcarbazole hydrochloride | 54-84-2 | MFCD00055609 | 99.5% | 376.9 g/mol | C20H25ClN2OS | 1 ML
SDP

Catalog No. 5000212363
Encompass_Preferred

Cinanserin hydrochloride is the hydrochloride salt of cinanserin, supplied for research use as a 10 mM solution in DMSO (1 mL) and as solid quantities. It functions as a 5-HT2 receptor antagonist in pharmacological and biochemical studies and is provided with supporting documentation for quality control.

  • Provided as 10 mM solution in DMSO (1 mL) and as solid pack sizes.
  • High purity suitable for biochemical and pharmacological assays.
  • Accompanied by a datasheet and certificate of analysis.
  • Used as a 5-HT2 receptor antagonist in research applications.
  • CAS number 54-84-2 for unambiguous chemical identification.
  • Molecular weight 376.9 g/mol.

Catalog No. 50-002-12363 Supplier Medchemexpress LLC Supplier No. HY10094310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cinanserin hydrochloride is the hydrochloride salt of cinanserin, supplied for research use as a 10 mM solution in DMSO (1 mL) and as solid quantities. It functions as a 5-HT2 receptor antagonist in pharmacological and biochemical studies and is provided with supporting documentation for quality control.

  • Provided as 10 mM solution in DMSO (1 mL) and as solid pack sizes.
  • High purity suitable for biochemical and pharmacological assays.
  • Accompanied by a datasheet and certificate of analysis.
  • Used as a 5-HT2 receptor antagonist in research applications.
  • CAS number 54-84-2 for unambiguous chemical identification.
  • Molecular weight 376.9 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.