Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Dimethylaminoethyl)-5-mercaptotetrazole 98.0+%, TCI America™
SDP

Catalog No. D203225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D203225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D203225G Supplier TCI America Supplier No. D203225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61607-68-9
Molecular Formula C5H11N5S
Molecular Weight (g/mol) 173.24
MDL Number MFCD00082923
InChI Key ODDAWJGQWOGBCX-UHFFFAOYSA-N
Synonym 1-(2-Dimethylaminoethyl)-1H-tetrazole-5-thiol
PubChem CID 3017268
IUPAC Name 1-[2-(dimethylamino)ethyl]-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
SMILES CN(C)CCN1NN=NC1=S

Specifications

Melting Point 205°C
Color White
Quantity 25 g
UN Number 1325
Formula Weight 173.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Dimethylaminoethyl)-5-mercaptotetrazole
EINECSNumber (5)-6307
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.