Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethylhydrazine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D074125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D074125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D074125G Supplier TCI America Supplier No. D074125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 306-37-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012522
InChI Key WLDRFJDGKMTPPV-UHFFFAOYSA-N
Synonym 1,2-dimethylhydrazine dihydrochloride, n,n'-dimethylhydrazine dihydrochloride, 1,2-dimethylhydrazinium dichloride, 1,2-dimethylhydrazine 2hcl, sym-dimethylhydrazine dihydrochloride, hydrazine, 1,2-dimethyl-, dihydrochloride, dimethylhydrazinium dichloride, ccris 260, 1,2-dimethylhydrazine.2hcl, hydrazine, 1,2-dimethyl-, hydrochloride 1:2
PubChem CID 9380
ChEBI CHEBI:73759
IUPAC Name dihydrogen 1,2-dimethylhydrazine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CNNC

Specifications

Melting Point 168°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 133.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethylhydrazine Dihydrochloride
RTECSNumber MV2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.