Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethylimidazole, 96+%

Catalog No. p-117949
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159441000 100 g Glass bottle
AC159445000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159441000 Supplier Thermo Scientific Chemicals Supplier No. 159441000
Only null left
Add to Cart

CAS: 1739-84-0 | C5H8N2 | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1739-84-0
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005294
InChI Key GIWQSPITLQVMSG-UHFFFAOYSA-N
Synonym 1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole
PubChem CID 15617
IUPAC Name 1,2-dimethylimidazole
SMILES CN1C=CN=C1C

Specifications

CAS Min % 96+%
Melting Point 37.0°C to 39.0°C
Color Colorless to Yellow
Density 1.0840g/mL
Boiling Point 205.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 66
Specific Gravity 1.084
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Viscosity 1.57 mPa.s (40°C)
Formula Weight 96.13
Percent Purity 96+%
Physical Form Low Melting Mass
Chemical Name or Material 1, 2-Dimethylimidazole, 96+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 217-101-2
TSCA TSCA
Recommended Storage may discolor during storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.