Learn More
Medchemexpress LLC 1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine | 78415-49-3 | >99.0% | 732.27 g·mol⁻¹ | C36H18D54NO8P | 5 MG
SDP
Catalog No. 5000299590
Encompass_Preferred
1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine is a perdeuterated phosphatidylcholine used as an isotopically labeled model lipid for membrane biophysics and structural studies such as neutron scattering and deuterium NMR.
  • Perdeuterated fatty acyl chains provide strong contrast for neutron scattering.
  • High isotopic and chemical purity (>99%).
  • Compatible with deuterium NMR and other spectroscopic techniques.
  • Used to form model membranes, liposomes, and supported bilayers.
  • Supplied as a dry powder suitable for dispersion and vesicle preparation.
Catalog No. 50-002-99590 Supplier Medchemexpress LLC Supplier No. HYW0199995MG
1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine is a perdeuterated phosphatidylcholine used as an isotopically labeled model lipid for membrane biophysics and structural studies such as neutron scattering and deuterium NMR.
  • Perdeuterated fatty acyl chains provide strong contrast for neutron scattering.
  • High isotopic and chemical purity (>99%).
  • Compatible with deuterium NMR and other spectroscopic techniques.
  • Used to form model membranes, liposomes, and supported bilayers.
  • Supplied as a dry powder suitable for dispersion and vesicle preparation.