Learn More
Medchemexpress LLC 1 2-Dimyristoyl-rac- 100mg
SDP
Catalog No. 5000206752
Encompass_Preferred
1 2-Dimyristoyl-rac-glycerol (C14 0) is a carboxylic acid ester glycerolipid and diglyceride with an additional myristoyl group that facilitates interactions between proteins and lipids Can be used as detergent or reagent It plays a role in cell biology experiments involving the metabolism and metabolic pathways of glycerolipids As a diglyceride this substance consists of two fatty acid chains covalently bonded in the 1 2-form to a glycerol molecule The diglyceride (DAG) study investigated the process by which DAG is depleted to inhibit fat accumulation Reagent grade for research use only
Catalog No. 50-002-06752 Supplier Medchemexpress LLC Supplier No. HYW127409100MG
1 2-Dimyristoyl-rac-glycerol (C14 0) is a carboxylic acid ester glycerolipid and diglyceride with an additional myristoyl group that facilitates interactions between proteins and lipids Can be used as detergent or reagent It plays a role in cell biology experiments involving the metabolism and metabolic pathways of glycerolipids As a diglyceride this substance consists of two fatty acid chains covalently bonded in the 1 2-form to a glycerol molecule The diglyceride (DAG) study investigated the process by which DAG is depleted to inhibit fat accumulation Reagent grade for research use only