Learn More
Medchemexpress LLC 1,2-dimyristoyl-rac-glycero-3-phosphocholine | 18656-38-7 | MFCD00066562 | 98.0% | 677.93 | C36H72NO8P | 100 MG
SDP
Catalog No. 5000282301
Encompass_Preferred
1,2-Dimyristoyl-rac-glycero-3-phosphocholine (DMPC) is a zwitterionic phospholipid used to model eukaryotic cell membranes. It is commonly employed to form bilayers and liposomes for membrane biophysics, formulation studies, and drug delivery research. Supplied as a defined-purity powder with storage recommendations for maintaining stability.
  • Zwitterionic phospholipid that mimics eukaryotic plasma membranes.
  • Forms stable bilayers and liposomes for membrane studies.
  • High purity (98.0%) suitable for analytical and preparative work.
  • Defined molecular weight and formula support reproducible experiments.
  • Recommended cold, moisture-controlled storage to preserve stability.
Catalog No. 50-002-82301 Supplier Medchemexpress LLC Supplier No. HYW142596100MG
1,2-Dimyristoyl-rac-glycero-3-phosphocholine (DMPC) is a zwitterionic phospholipid used to model eukaryotic cell membranes. It is commonly employed to form bilayers and liposomes for membrane biophysics, formulation studies, and drug delivery research. Supplied as a defined-purity powder with storage recommendations for maintaining stability.
  • Zwitterionic phospholipid that mimics eukaryotic plasma membranes.
  • Forms stable bilayers and liposomes for membrane studies.
  • High purity (98.0%) suitable for analytical and preparative work.
  • Defined molecular weight and formula support reproducible experiments.
  • Recommended cold, moisture-controlled storage to preserve stability.