Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1 2-Dimyristoyl-sn-g 100mg | 105405-50-3 | 100 MG
SDP

Catalog No. 5000308014
Encompass_Preferred

1,2-Dimyristoyl-sn-glycero-3-phospho-L-serine sodium is the sodium salt of dimyristoyl phosphatidylserine, a negatively charged phospholipid used in membrane biophysics and liposome formulation. It is supplied as a white to off-white solid and is commonly used to incorporate phosphatidylserine into membrane model systems and liposomes.

  • 98.0% purity, suitable for research applications.
  • Molecular weight 701.84 g/mol and formula C34H65NNaO10P.
  • White to off-white solid appearance.
  • Recommended storage at -20°C, sealed and protected from moisture.
  • Suitable for membrane model systems and liposome preparation.

Catalog No. 50-003-08014 Supplier Medchemexpress LLC Supplier No. HY115435100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dimyristoyl-sn-glycero-3-phospho-L-serine sodium is the sodium salt of dimyristoyl phosphatidylserine, a negatively charged phospholipid used in membrane biophysics and liposome formulation. It is supplied as a white to off-white solid and is commonly used to incorporate phosphatidylserine into membrane model systems and liposomes.

  • 98.0% purity, suitable for research applications.
  • Molecular weight 701.84 g/mol and formula C34H65NNaO10P.
  • White to off-white solid appearance.
  • Recommended storage at -20°C, sealed and protected from moisture.
  • Suitable for membrane model systems and liposome preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.