Learn More
Medchemexpress LLC 1 2-Dimyristoyl-sn-g 10mg | 105405-50-3 | 701.84 | C34H65NNaO10P | 10 MG
SDP
Catalog No. 5000308015
Encompass_Preferred
1,2-dimyristoyl-sn-glycero-3-phospho-L-serine sodium is an anionic phospholipid bearing two myristoyl (14:0) acyl chains. It is used as a phosphatidylserine component in liposome formulation and membrane-mimetic studies to investigate membrane charge and protein-lipid interactions.
  • Anionic phospholipid with myristic acid (14:0) chains.
  • Suitable for liposome preparation and membrane model studies.
  • Sodium salt form; chemical formula C34H65NNaO10P, molecular weight 701.84.
  • Typical purity 98.0% by NMR based on certificate of analysis.
  • Available in research-scale quantities for formulation and biophysical assays.
Catalog No. 50-003-08015 Supplier Medchemexpress LLC Supplier No. HY11543510MG
1,2-dimyristoyl-sn-glycero-3-phospho-L-serine sodium is an anionic phospholipid bearing two myristoyl (14:0) acyl chains. It is used as a phosphatidylserine component in liposome formulation and membrane-mimetic studies to investigate membrane charge and protein-lipid interactions.
  • Anionic phospholipid with myristic acid (14:0) chains.
  • Suitable for liposome preparation and membrane model studies.
  • Sodium salt form; chemical formula C34H65NNaO10P, molecular weight 701.84.
  • Typical purity 98.0% by NMR based on certificate of analysis.
  • Available in research-scale quantities for formulation and biophysical assays.