Learn More
Medchemexpress LLC 1 2-Dimyristoyl-sn-g 25mg | 105405-50-3 | 701.84 | C34H65NNaO10P | 25 MG
SDP
Catalog No. 5000308017
Encompass_Preferred
An anionic phospholipid (sodium salt of phosphatidylserine) commonly used in liposome and membrane formulation and membrane biology research. It provides anionic surface charge for model membranes and is supplied with high listed purity suitable for research use.
  • Used in liposome and membrane formulation and stability studies.
  • Provides anionic charge as the sodium salt form for membrane modeling.
  • Molecular formula C34H65NNaO10P and molecular weight 701.84.
  • Listed purity 98.0% appropriate for research applications.
Catalog No. 50-003-08017 Supplier Medchemexpress LLC Supplier No. HY11543525MG
An anionic phospholipid (sodium salt of phosphatidylserine) commonly used in liposome and membrane formulation and membrane biology research. It provides anionic surface charge for model membranes and is supplied with high listed purity suitable for research use.
  • Used in liposome and membrane formulation and stability studies.
  • Provides anionic charge as the sodium salt form for membrane modeling.
  • Molecular formula C34H65NNaO10P and molecular weight 701.84.
  • Listed purity 98.0% appropriate for research applications.