Learn More
Medchemexpress LLC 1 2-Dimyristoyl-sn-g 50mg | 105405-50-3 | 50 MG
SDP
Catalog No. 5000228757
Encompass_Preferred
1,2-dimyristoyl-sn-glycero-3-phospho-L-serine sodium is an anionic phospholipid (sodium salt) with myristoyl (14:0) fatty acid tails used in lipid research and formulation work such as liposome preparation and membrane model studies.
  • Used for liposome preparation and membrane model studies.
  • Contains defined 14:0 fatty acid tails for consistent biophysical properties.
  • High research-grade purity (98.0%) suitable for analytical and formulation work.
  • Available in small laboratory pack sizes for efficient use.
Catalog No. 50-002-28757 Supplier Medchemexpress LLC Supplier No. HY11543550MG
1,2-dimyristoyl-sn-glycero-3-phospho-L-serine sodium is an anionic phospholipid (sodium salt) with myristoyl (14:0) fatty acid tails used in lipid research and formulation work such as liposome preparation and membrane model studies.
  • Used for liposome preparation and membrane model studies.
  • Contains defined 14:0 fatty acid tails for consistent biophysical properties.
  • High research-grade purity (98.0%) suitable for analytical and formulation work.
  • Available in small laboratory pack sizes for efficient use.