Learn More
Avanti Polar Lipids 14:0 PC (DMPC) | 18194-24-6 | MFCD00036901 | 25mg
Catalog No. NC1371620
Encompass
14:0 PC (DMPC) | Putity: >99% | MW: 677.95 | 18194-24-6 | MFCD00036901 | 25mg
Catalog No. NC1371620 Supplier Avanti Polar Lipids Supplier No. 850345P25MG
14:0 PC (DMPC) | Putity: >99% | MW: 677.95 | 18194-24-6 | MFCD00036901 | 25mg