Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dimyristoyl-sn-glycero-3-phosphate monosodium | 80724-31-8 | 98.0% | 614.77 g·mol⁻¹ | C31H60NaO8P | 25 MG
SDP

Catalog No. 5000209199
Encompass_Preferred

1,2-dimyristoyl-sn-glycero-3-phosphate monosodium is a saturated phospholipid with two myristoyl (14:0) acyl chains at the sn-1 and sn-2 positions. Supplied as a white to off-white solid, it is used to prepare micelles, liposomes, and model membrane systems for biochemical and biophysical research. Data sheet, certificate of analysis, and safety data sheet are available for handling and formulation guidance.

  • White to off-white solid suitable for membrane research.
  • High purity (98.0%) appropriate for formulation of liposomes and micelles.
  • Molecular weight 614.77 g·mol⁻¹ and formula C31H60NaO8P for stoichiometric calculations.
  • Solvent-specific solubility information available in the product documentation.
  • Store sealed, away from moisture and light; in solution store at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-09199 Supplier Medchemexpress LLC Supplier No. 25MGHYW040185
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-dimyristoyl-sn-glycero-3-phosphate monosodium is a saturated phospholipid with two myristoyl (14:0) acyl chains at the sn-1 and sn-2 positions. Supplied as a white to off-white solid, it is used to prepare micelles, liposomes, and model membrane systems for biochemical and biophysical research. Data sheet, certificate of analysis, and safety data sheet are available for handling and formulation guidance.

  • White to off-white solid suitable for membrane research.
  • High purity (98.0%) appropriate for formulation of liposomes and micelles.
  • Molecular weight 614.77 g·mol⁻¹ and formula C31H60NaO8P for stoichiometric calculations.
  • Solvent-specific solubility information available in the product documentation.
  • Store sealed, away from moisture and light; in solution store at -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.