Learn More
Medchemexpress LLC 1,2-dimyristoyl-sn-glycero-3-phosphate monosodium | 80724-31-8 | MFCD00153886 | 98.0% | 614.77 g/mol | C31H60NaO8P | 100 MG
SDP
Catalog No. 5000209235
Encompass_Preferred
1,2-Dimyristoyl-sn-glycero-3-phosphate monosodium is a synthetic phospholipid (phosphatidic acid) in which myristic acid (14:0) is esterified at both the sn-1 and sn-2 positions. It is supplied as the sodium salt and is commonly used to prepare liposomes, micelles, and other model membrane systems for biochemical and biophysical research.
  • Synthetic phospholipid with tetradecanoyl (14:0) chains at sn-1 and sn-2 positions.
  • Useful for generation of micelles, liposomes, and artificial membranes.
  • Supplied as a solid, white to off-white, sodium salt.
  • Purity typically 98.0% as provided by vendor specifications.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
Catalog No. 50-002-09235 Supplier Medchemexpress LLC Supplier No. HYW040185100MG
1,2-Dimyristoyl-sn-glycero-3-phosphate monosodium is a synthetic phospholipid (phosphatidic acid) in which myristic acid (14:0) is esterified at both the sn-1 and sn-2 positions. It is supplied as the sodium salt and is commonly used to prepare liposomes, micelles, and other model membrane systems for biochemical and biophysical research.
  • Synthetic phospholipid with tetradecanoyl (14:0) chains at sn-1 and sn-2 positions.
  • Useful for generation of micelles, liposomes, and artificial membranes.
  • Supplied as a solid, white to off-white, sodium salt.
  • Purity typically 98.0% as provided by vendor specifications.
  • Store sealed at 4°C; in solvent, store at -80°C up to 6 months or -20°C up to 1 month.