Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine 97.0+%, TCI America™
SDP

Catalog No. D4249250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4249250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4249250MG Supplier TCI America Supplier No. D4249250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 998-07-2
Molecular Formula C33H66NO8P
Molecular Weight (g/mol) 635.864
MDL Number MFCD00036775
InChI Key NEZDNQCXEZDCBI-WJOKGBTCSA-N
Synonym 1,2-Ditetradecanoyl-sn-glycero-3-phosphoethanolamine, 2-Aminoethyl (R)-2,3-Bis(tetradecanoyloxy)propyl Hydrogen Phosphate, L-DMPE
PubChem CID 9852308
IUPAC Name [(2R)-3-[2-aminoethoxy(hydroxy)phosphoryl]oxy-2-tetradecanoyloxypropyl] tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCCN)OC(=O)CCCCCCCCCCCCC

Specifications

Melting Point 193°C
Color White
Quantity 250 mg
Formula Weight 635.86
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.