Learn More
Medchemexpress LLC 1,2-Dimyristoyl-sn-glycerol | 60562-16-5 | 99.96% | 100 MG
SDP
Catalog No. 5000342017
Encompass_Preferred
1,2-Dimyristoyl-sn-glycerol is a saturated diacylglycerol that acts as a weak second messenger for the activation of Protein Kinase C (PKC). This product is intended for research use only.
  • Saturated diacylglycerol compound
  • Weak second messenger for protein kinase C activation
  • Suitable for biochemical assay research
  • Applicable in drug delivery studies
  • Used in research for infection, inflammation, and digestive system diseases
Catalog No. 50-003-42017 Supplier Medchemexpress LLC Supplier No. HY128468100MG
1,2-Dimyristoyl-sn-glycerol is a saturated diacylglycerol that acts as a weak second messenger for the activation of Protein Kinase C (PKC). This product is intended for research use only.
  • Saturated diacylglycerol compound
  • Weak second messenger for protein kinase C activation
  • Suitable for biochemical assay research
  • Applicable in drug delivery studies
  • Used in research for infection, inflammation, and digestive system diseases