Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2'-Dinaphthylamine (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. D29885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29885G 5 g
D29881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29885G Supplier TCI America Supplier No. D29885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4669-06-1
Molecular Formula C20H15N
Molecular Weight (g/mol) 269.35
InChI Key UNJZLNFHHINVOB-UHFFFAOYSA-N
PubChem CID 257254
IUPAC Name N-naphthalen-1-ylnaphthalen-2-amine
SMILES C1=CC=C2C=C(C=CC2=C1)NC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 5 g
Synonym 1,2′-Iminodinaphthalene
Formula Weight 269.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2′-Dinaphthylamine (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.