Learn More
Enzo Life Sciences 1,2-Dioctanoyl-sn-glycerol (8:0) (20mg). CAS: 60514-48-9
SDP
Supplier:  Enzo Life Sciences BMLDG1120020
Encompass_Preferred
Potent activator of protein kinase C (PKC). Membrane permeable but non-physiologic DAG. Induces a discrete but transient translocation of protein kinase C. Alternative name: DOG. Purity: ≥96% (TLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -80°C.
Catalog No. 50-201-0507