Learn More
Avanti Polar Lipids 1,2-dioctanoyl-sn-glycero-3-phospho-(1'-myo-inositol) (ammonium salt), 2260795-76-2, 500 ug
Supplier:  Avanti Polar Lipids 850181P500UG
Encompass
Molecular Formula: C25H50NO13P, Formula Weight: 603.637, Purity >99%. Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.
Catalog No. NC0816085