Learn More
Avanti Polar Lipids 1, 2-Dioleoyl-sn-Glycero-3-Phosphocholine (DOPC), Hygroscopic as Chloroform, 200ml
Supplier:  Avanti Polar Lipids 850375C200MG
Encompass
1, 2-Dioleoyl-sn-Glycero-3-Phosphocholine (DOPC), Hygroscopic as Chloroform, C 44 H 84 NO8P, Percent composition: C 67.23% H 10.77% N 1.78% O 16.28% P 3.94%, 200mg
Catalog No. NC9276550