Learn More
Medchemexpress LLC 1,2-Dioleoyl-3-linoleoyl-rac-glycerol | 2190-20-7 | 50 MG
SDP
Catalog No. 5000418156
Encompass_Preferred
1,2-Dioleoyl-3-linoleoyl-rac-glycerol (OOL) is a liquid triacylglycerol that serves as an endogenous metabolite. It is characterized by containing oleic acid at the sn-1 and sn-2 positions and linoleic acid at the sn-3 position. This compound is naturally found in a variety of seed and vegetable oils, such as pumpkin seed, olive, and sesame oils. Primarily, it is utilized as a laboratory chemical for research purposes and in the manufacturing of substances, playing a role in metabolic enzyme and protease pathways.
  • Triacylglycerol composed of oleic acid and linoleic acid
  • Found in various seed and vegetable oils
  • Used as a laboratory chemical and for substance manufacturing
  • Involved in metabolic enzyme/protease pathways
  • Appears as a liquid
  • Functions as an endogenous metabolite
Catalog No. 50-004-18156 Supplier Medchemexpress LLC Supplier No. HYN795350MG
1,2-Dioleoyl-3-linoleoyl-rac-glycerol (OOL) is a liquid triacylglycerol that serves as an endogenous metabolite. It is characterized by containing oleic acid at the sn-1 and sn-2 positions and linoleic acid at the sn-3 position. This compound is naturally found in a variety of seed and vegetable oils, such as pumpkin seed, olive, and sesame oils. Primarily, it is utilized as a laboratory chemical for research purposes and in the manufacturing of substances, playing a role in metabolic enzyme and protease pathways.
  • Triacylglycerol composed of oleic acid and linoleic acid
  • Found in various seed and vegetable oils
  • Used as a laboratory chemical and for substance manufacturing
  • Involved in metabolic enzyme/protease pathways
  • Appears as a liquid
  • Functions as an endogenous metabolite