Learn More
TARGETMOL CHEMICALS INC 1 2-Dioleoyl-sn-glycero-3-phos
SDP
Catalog No. 502879009
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1,2-Dioleoyl-sn-glycero-3-phospho-L-serine sodium (1,2-Dioleoyl-sn-glycero-3-PS (sodium salt)) can be used in lipid mixtures with DOPC and DOPE as effective nontoxic and nonviral DNA vectors. It has been utilized in the formation of unilamellar vesicles to study membrane structure curvature and in supported lipid bilayers to examine the impact of various support materials on lipid redistribution between membrane leaflets. Purity 98.93%
Catalog No. 50-287-9009 Supplier TARGETMOL CHEMICALS INC Supplier No. T3560650MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 1,2-Dioleoyl-sn-glycero-3-phospho-L-serine sodium (1,2-Dioleoyl-sn-glycero-3-PS (sodium salt)) can be used in lipid mixtures with DOPC and DOPE as effective nontoxic and nonviral DNA vectors. It has been utilized in the formation of unilamellar vesicles to study membrane structure curvature and in supported lipid bilayers to examine the impact of various support materials on lipid redistribution between membrane leaflets. Purity 98.93%